Learn More
Cayman Chemical 4'-O-methyl Quercetin, Purity: ≥95%, CAS: 603-61-2, 10 mg
SDP
Catalog No. 502986023
Encompass_Preferred
A flavonoid from C. ordata with anticancer and antiplasmodial activity; inhibits the viability of HL-60, U937, MOLT-3, Raji, K562, MCF-7, SK-MEL-1, and A549 human tumor cell lines with IC50 values ranging from 5.5-24.1 μM; induces G2-M arrest and inhibits tubulin polymerization in vitro; inhibits breast cancer resistance protein (BCRP/ABCG2; IC50 = 0.040 μM in a vesicular transport assay); reduces in vitro proliferation of chloroquine-resistant P. falciparum (IC50 = 4.8 μM)
Catalog No. 50-298-6023 Supplier Cayman Chemical Supplier No. 2240610 MG
A flavonoid from C. ordata with anticancer and antiplasmodial activity; inhibits the viability of HL-60, U937, MOLT-3, Raji, K562, MCF-7, SK-MEL-1, and A549 human tumor cell lines with IC50 values ranging from 5.5-24.1 μM; induces G2-M arrest and inhibits tubulin polymerization in vitro; inhibits breast cancer resistance protein (BCRP/ABCG2; IC50 = 0.040 μM in a vesicular transport assay); reduces in vitro proliferation of chloroquine-resistant P. falciparum (IC50 = 4.8 μM)