Learn More
Medchemexpress LLC HY-N1910 5mg Medchemexpress, 4'-O-Methylbavachalcone CAS:20784-60-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN19105MG
Encompass_Preferred
Medchemexpress, HY-N1910 5mg 4'-O-Methylbavachalcone CAS:20784-60-5 4'-O-Methylbavachalcone is a chalcone isolated from Psoralea corylifolia, inhibits severe acute respiratory syndrome coronavirus (SARS-CoV) papain-like protease (PLpro) activity, with an IC50 of 10.1 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0350