Learn More
Medchemexpress LLC 4-O-methyldopamine hydrochloride | 645-33-0 | MFCD00012896 | 99.5% | 203.67 g/mol | C9H14ClNO2 | 250 MG
SDP
Catalog No. 5000209270
Encompass_Preferred
4-O-Methyldopamine hydrochloride is the hydrochloride salt of a catecholamine compound used as a research reagent; it is reported to inhibit dihydropteridine reductase and is supplied as a high-purity solid for biochemical and pharmacological studies.
  • Hydrochloride salt form for improved solubility and stability.
  • Reported inhibitor of dihydropteridine reductase for enzyme studies.
  • High purity: 99.46%.
  • Molecular weight: 203.67 g/mol; CAS number: 645-33-0.
  • Supplied as a solid powder suitable for analytical and preparative work.
  • Available in multiple pack sizes, including 250 mg, to support varying experimental scales.
Catalog No. 50-002-09270 Supplier Medchemexpress LLC Supplier No. 250MGHYW041498
4-O-Methyldopamine hydrochloride is the hydrochloride salt of a catecholamine compound used as a research reagent; it is reported to inhibit dihydropteridine reductase and is supplied as a high-purity solid for biochemical and pharmacological studies.
  • Hydrochloride salt form for improved solubility and stability.
  • Reported inhibitor of dihydropteridine reductase for enzyme studies.
  • High purity: 99.46%.
  • Molecular weight: 203.67 g/mol; CAS number: 645-33-0.
  • Supplied as a solid powder suitable for analytical and preparative work.
  • Available in multiple pack sizes, including 250 mg, to support varying experimental scales.