Learn More
Cayman Chemical 4'-O-Methylhonokiol, Purity: ≥95%, CAS: 68592-15-4, 10 mg
SDP
Catalog No. 502954062
Encompass_Preferred
A phenol with diverse biological activities; active against the bacteria B. subtilis, S. aureus, and M. smegmatis (MIC = 2.5 µg/ml for all) and the fungus T. mentagrophytes (MIC = 1.25 µg/ml); inhibits LPS-induced activation of NF-kB and production of NO in RAW 264.7 macrophages at 10 and 30 µM; reduces carrageenan-induced paw edema in mice at 20 and 200 mg/kg; reduces neuronal apoptosis and escape latency in the Morris water maze in a mouse model of Aβ42-induced Alzheimer's disease at 1 mg/kg; reduces plasma cholesterol and triglyceride levels, ameliorates hepatic steatosis, and improves insulin resistance in a mouse model of high-fat diet-induced obesity
Catalog No. 50-295-4062 Supplier Cayman Chemical Supplier No. 1108910 MG
A phenol with diverse biological activities; active against the bacteria B. subtilis, S. aureus, and M. smegmatis (MIC = 2.5 µg/ml for all) and the fungus T. mentagrophytes (MIC = 1.25 µg/ml); inhibits LPS-induced activation of NF-kB and production of NO in RAW 264.7 macrophages at 10 and 30 µM; reduces carrageenan-induced paw edema in mice at 20 and 200 mg/kg; reduces neuronal apoptosis and escape latency in the Morris water maze in a mouse model of Aβ42-induced Alzheimer's disease at 1 mg/kg; reduces plasma cholesterol and triglyceride levels, ameliorates hepatic steatosis, and improves insulin resistance in a mouse model of high-fat diet-induced obesity