Learn More
Cayman Chemical 4'-O-Methylresveratrol, Purity: ≥98%, CAS: 33626-08-3, 50 mg
SDP
Catalog No. 502940558
Encompass_Preferred
A stilbene with diverse biological activities; inhibits growth of A549, SKOV3, SK-MEL-2, XF498, and HCT15 cancer cells (IC50s = 3.8, 5.7, 3.2, 5.6, and 6.7 µg/ml, respectively); inhibits arachidonic acid- or collagen-induced rabbit platelet aggregation from 0.05-0.2 mM; scavenges free radicals in a TEAC assay; suppresses RANKL-induced osteoclastic differentiation of murine BMDMs; reduces LPS-induced bone loss in mice at 50 mg/kg
Catalog No. 50-294-0558 Supplier Cayman Chemical Supplier No. 2910650 MG
A stilbene with diverse biological activities; inhibits growth of A549, SKOV3, SK-MEL-2, XF498, and HCT15 cancer cells (IC50s = 3.8, 5.7, 3.2, 5.6, and 6.7 µg/ml, respectively); inhibits arachidonic acid- or collagen-induced rabbit platelet aggregation from 0.05-0.2 mM; scavenges free radicals in a TEAC assay; suppresses RANKL-induced osteoclastic differentiation of murine BMDMs; reduces LPS-induced bone loss in mice at 50 mg/kg