Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4'-Octyl-4-biphenylcarbonitrile

Catalog No. 3386801G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-868-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 33-868-01G Supplier Sigma Aldrich Supplier No. 3386801G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4G-octyl-4-biphenylcarbonitrile (8CB) is a low molar mass liquid crystal (LC), which shows different liquid crystalline phases depending on temperature conditions. Under normal temperature conditions 8CB exists only as a bulk smectic-A phase. LCs are responsive to subtle changes in temperature, shear, electric (or magnetic) fields, and the structure of solid surfaces with which they are in contact.Monolayer films of the LCs deposited onto highly ordered pyrolytic graphite, these self-assembled films display a wide variety of structural forms.

Specifications

CAS 52709-84-9
Molecular Formula C21H25N
Linear Formula CH3(CH2)7C6H4C6H4CN
Quantity 1 g
MDL Number MFCD00075146
Synonym 4 -n -Octyl-4-cyanobiphenyl; 4 -Octyl-4-biphenylcarbonitrile; 4 -Octyl-4-cyanobiphenyl; 4 -Octyl[1,1 -biphenyl]-4-carbonitrile; 4-n -Octyl-4 -cyanobiphenyl; 4-Cyano-4 -n -octylbiphenyl; 4-Cyano-4 -octyl-1,1 -biphenyl; 4-Cyano-4 -octylbiphenyl; 4-Octyl-4 -cyanobiphenyl; p -n -Octyl-p -cyanobiphenyl; p -Cyano-p -octylbiphenyl; p -Octyl-p -cyanobiphenyl
Molecular Weight (g/mol) 291.43
Percent Purity 98%
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.