Learn More
Selleck Chemical LLC 4-Octyl Itaconate
SDP
Catalog No. 5000003627
Encompass_Preferred
4-Octyl Itaconate activates Nrf2 signaling to inhibit the production of pro-inflammatory cytokines in human macrophages and systemic lupus erythematosus (SLE) patient-derived PBMCs
Catalog No. 50-000-03627 Supplier Selleck Chemical LLC Supplier No. S59291G
4-Octyl Itaconate activates Nrf2 signaling to inhibit the production of pro-inflammatory cytokines in human macrophages and systemic lupus erythematosus (SLE) patient-derived PBMCs