Learn More
Medchemexpress LLC HY-112675 200mg , 4-Octyl Itaconate CAS:3133-16-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY112675200MG
Encompass_Preferred
HY-112675 200mg 4-Octyl Itaconate CAS: 3133-16-2 4-Octyl Itaconate is a cell-permeable Itaconate derivative. Itaconate is an anti-inflammatory metabolite that activates Nrf2 via alkylation of KEAP1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9458