Learn More
Cayman Chemical 4-octyl Itaconate, Purity: ≥98% (mixture of tautomers), CAS: 3133-16-2, 25 mg
SDP
Catalog No. 502934211
Encompass_Preferred
A prodrug form of itaconate; hydrolyzed to itaconate by esterases in C2C12 cells and LPS-activated mouse macrophages; increases Nrf2 protein levels and expression of Nrf2 target genes and enhances LPS-induced Nrf2 stabilization in mouse macrophages; decreases serum levels of IL-1β and TNF-α, increases Nrf2 protein expression, and protects against LPS-induced lethality in wild-type, but not Nrf2 knockout, mice at 50 mg/kg
Catalog No. 50-293-4211 Supplier Cayman Chemical Supplier No. 2537425 MG
A prodrug form of itaconate; hydrolyzed to itaconate by esterases in C2C12 cells and LPS-activated mouse macrophages; increases Nrf2 protein levels and expression of Nrf2 target genes and enhances LPS-induced Nrf2 stabilization in mouse macrophages; decreases serum levels of IL-1β and TNF-α, increases Nrf2 protein expression, and protects against LPS-induced lethality in wild-type, but not Nrf2 knockout, mice at 50 mg/kg