Learn More
Medchemexpress LLC 4-Octyl itaconate | 3133-16-2 | 99.9% | 242.31 | 1 ML
SDP
Catalog No. 5000437884
Encompass_Preferred
4-Octyl itaconate (4-OI) is a cell-permeable itaconate derivative. Itaconate is an anti-inflammatory metabolite that activates Nrf2 via alkylation of KEAP1, playing a protective role against lipopolysaccharide-induced lethality in vivo by decreasing cytokine production.
  • Cell-permeable
  • Anti-inflammatory metabolite
  • Nrf2 Activator
  • Protective against LPS-induced lethality in vivo
Catalog No. 50-004-37884 Supplier Medchemexpress LLC Supplier No. HY11267510MM/1ML
4-Octyl itaconate (4-OI) is a cell-permeable itaconate derivative. Itaconate is an anti-inflammatory metabolite that activates Nrf2 via alkylation of KEAP1, playing a protective role against lipopolysaccharide-induced lethality in vivo by decreasing cytokine production.
  • Cell-permeable
  • Anti-inflammatory metabolite
  • Nrf2 Activator
  • Protective against LPS-induced lethality in vivo