Learn More
Medchemexpress LLC 4-Octyl itaconate | 3133-16-2 | 99.9% | 242.31 | 10 MG
SDP
Catalog No. 5000436990
Encompass_Preferred
4-Octyl Itaconate is a cell-permeable itaconate derivative and anti-inflammatory metabolite that activates Nrf2 via alkylation of KEAP1. It is protective against lipopolysaccharide-induced lethality in vivo and decreases cytokine production.
  • Cell-permeable itaconate derivative
  • Activates Nrf2 via alkylation of KEAP1
  • Protective against lipopolysaccharide-induced lethality in vivo
  • Decreases cytokine production
  • For research use only
Catalog No. 50-004-36990 Supplier Medchemexpress LLC Supplier No. HY11267510MG
4-Octyl Itaconate is a cell-permeable itaconate derivative and anti-inflammatory metabolite that activates Nrf2 via alkylation of KEAP1. It is protective against lipopolysaccharide-induced lethality in vivo and decreases cytokine production.
  • Cell-permeable itaconate derivative
  • Activates Nrf2 via alkylation of KEAP1
  • Protective against lipopolysaccharide-induced lethality in vivo
  • Decreases cytokine production
  • For research use only