Learn More
Medchemexpress LLC HY-112675 100mg Medchemexpress, 4-Octyl Itaconate CAS:3133-16-2 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY112675100MG
Encompass_Preferred
Medchemexpress,HY-112675 100mg 4-Octyl Itaconate CAS:3133-16-2 Formula:C13H22O4 4-Octyl Itaconate is a cell-permeable Itaconate derivative. Itaconate is an anti-inflammatory metabolite that activates Nrf2 via alkylation of KEAP1. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7621