Learn More
Selleck Chemical LLC 4-Octyl Itaconate 5mg 3133-16-2
SDP
Supplier:  Selleck Chemical LLC S59295MG
Encompass_Preferred
4-Octyl Itaconate activates Nrf2 signaling to inhibit the production of pro-inflammatory cytokines in human macrophages and systemic lupus erythematosus (SLE) patient-derived PBMCs. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8331