Learn More
Cayman Chemical 4-octynyl Itaconate 10mg
SDP
Catalog No. 502937821
Encompass_Preferred
An alkyne derivative of 4-octyl itaconate; can be used for click chemistry applications
Catalog No. 50-293-7821 Supplier Cayman Chemical Supplier No. 2845510 MG
An alkyne derivative of 4-octyl itaconate; can be used for click chemistry applications