Learn More
Julabo USA Inc Flat bath cover for PURA 10 with 4 openings, 92 mm dia. and sets of rings.
SDP
Catalog No. 501904638
Encompass_Preferred
Flat stainless steel covers for JULABO PURA water baths. Lids include concentric ring openings for manual adjustement of opening size. For use with tall flasks or vessels to minimize water evaporation in the bath tank.
Catalog No. 50-190-4638 Supplier Julabo USA Inc Supplier No. 9970570
Flat stainless steel covers for JULABO PURA water baths. Lids include concentric ring openings for manual adjustement of opening size. For use with tall flasks or vessels to minimize water evaporation in the bath tank.