Learn More
Chemglass Life Sciences 4-Outlet Power Strip
SDP
Catalog No. 50872322
Encompass_Preferred
4-Outlet Power Strip. A unique 4-outlet power strip having individual lighted switches and breaker. Supplied complete with mounting arm for attachment to any Chemglass support frame. A universal Kwik Klamp II is needed for mounting to support frame, see CG-9253.
Catalog No. 50-872-322 Supplier Chemglass Life Sciences Supplier No. CG196580
4-Outlet Power Strip. A unique 4-outlet power strip having individual lighted switches and breaker. Supplied complete with mounting arm for attachment to any Chemglass support frame. A universal Kwik Klamp II is needed for mounting to support frame, see CG-9253.