Learn More
Medchemexpress LLC 4-Oxazolecarboxamide, N-[6-[(2-methoxyethyl)methylamino]-3-pyridinyl]-2-phenyl-5-(trifluorom | 939375-07-2 | 99.9% | 420.39 | 100 MG
SDP
Catalog No. 5000393984
Encompass_Preferred
DGAT1-IN-3 is a potent, selective, and orally bioavailable inhibitor of DGAT-1. It can be used for research on obesity, dyslipidemia, and metabolic syndrome.
  • Potent, selective, and orally bioavailable inhibitor of DGAT-1.
  • Inhibits human DGAT-1 with an IC50 of 38 nM.
  • Inhibits rat DGAT-1 with an IC50 of 120 nM.
  • Supports research into obesity.
  • Supports research into dyslipidemia.
  • Supports research into metabolic syndrome.
  • Reduces cumulative body weight gain in DIO rats.
  • Reduces weight gain and plasma triglycerides.
  • Improves lipid profiles.
Catalog No. 50-003-93984 Supplier Medchemexpress LLC Supplier No. HY16434100MG
DGAT1-IN-3 is a potent, selective, and orally bioavailable inhibitor of DGAT-1. It can be used for research on obesity, dyslipidemia, and metabolic syndrome.
  • Potent, selective, and orally bioavailable inhibitor of DGAT-1.
  • Inhibits human DGAT-1 with an IC50 of 38 nM.
  • Inhibits rat DGAT-1 with an IC50 of 120 nM.
  • Supports research into obesity.
  • Supports research into dyslipidemia.
  • Supports research into metabolic syndrome.
  • Reduces cumulative body weight gain in DIO rats.
  • Reduces weight gain and plasma triglycerides.
  • Improves lipid profiles.