Learn More
Medchemexpress LLC 4-Oxazolecarboxamide, N-[6-[(2-methoxyethyl)methylamino]-3-pyridinyl]-2-phenyl-5-(trifluoromethyl)- | 939375-07-2 | 99.9% | 420.39 | 5 MG
SDP
Catalog No. 5000394191
Encompass_Preferred
DGAT1-IN-3 is a potent, selective, and orally bioavailable inhibitor of DGAT-1, with IC50s of 38 nM for human DGAT-1 and 120 nM for rat DGAT-1. It can be used for research on obesity, dyslipidemia, and metabolic syndrome.
  • Potent, selective, and orally bioavailable inhibitor of DGAT-1.
  • IC50s: 38 nM (human DGAT-1); 120 nM (rat DGAT-1).
  • Reduces weight gain and plasma triglycerides, and improves lipid profile in animal models.
  • Exhibits good oral bioavailability (77%) and a maximum exposure level in plasma (Cmax) of 24 μM (50 mg/kg; p.o).
  • Exhibits terminal elimination half-lives (1.95 h) and low clearance (13.5 mL/min/kg) (5 mg/kg; i.v).
  • Appearance: solid, light yellow to yellow color.
Catalog No. 50-003-94191 Supplier Medchemexpress LLC Supplier No. HY164345MG
DGAT1-IN-3 is a potent, selective, and orally bioavailable inhibitor of DGAT-1, with IC50s of 38 nM for human DGAT-1 and 120 nM for rat DGAT-1. It can be used for research on obesity, dyslipidemia, and metabolic syndrome.
  • Potent, selective, and orally bioavailable inhibitor of DGAT-1.
  • IC50s: 38 nM (human DGAT-1); 120 nM (rat DGAT-1).
  • Reduces weight gain and plasma triglycerides, and improves lipid profile in animal models.
  • Exhibits good oral bioavailability (77%) and a maximum exposure level in plasma (Cmax) of 24 μM (50 mg/kg; p.o).
  • Exhibits terminal elimination half-lives (1.95 h) and low clearance (13.5 mL/min/kg) (5 mg/kg; i.v).
  • Appearance: solid, light yellow to yellow color.