Learn More
Medchemexpress LLC 4-oxazolecarboxamide, N-[6-[(2-methoxyethyl)methylamino]-3-pyridinyl]-2-phenyl-5-(trifluoromethyl)- | 939375-07-2 | 99.9% | 420.39 g/mol | C20H19F3N4O3 | 10 MG
SDP
Catalog No. 5000330265
Encompass_Preferred
DGAT1-IN-3 is a potent, selective, and orally bioavailable small-molecule inhibitor of diacylglycerol O-acyltransferase 1 (DGAT-1) intended as a research reagent for studies of lipid metabolism, obesity, and related metabolic disorders.
  • Potent DGAT-1 inhibition with human IC50 38 nM and rat IC50 120 nM.
  • High purity as verified by LCMS (99.92%).
  • Molecular formula C20H19F3N4O3 and molecular weight 420.39 g/mol.
  • Suitable for in vitro and in vivo pharmacology studies of lipid metabolism.
  • Available in small research quantities (1 mg, 5 mg, 10 mg).
  • Structural formula and quality documentation provided in the product datasheet and COA.
Catalog No. 50-003-30265 Supplier Medchemexpress LLC Supplier No. HY1643410MG
DGAT1-IN-3 is a potent, selective, and orally bioavailable small-molecule inhibitor of diacylglycerol O-acyltransferase 1 (DGAT-1) intended as a research reagent for studies of lipid metabolism, obesity, and related metabolic disorders.
  • Potent DGAT-1 inhibition with human IC50 38 nM and rat IC50 120 nM.
  • High purity as verified by LCMS (99.92%).
  • Molecular formula C20H19F3N4O3 and molecular weight 420.39 g/mol.
  • Suitable for in vitro and in vivo pharmacology studies of lipid metabolism.
  • Available in small research quantities (1 mg, 5 mg, 10 mg).
  • Structural formula and quality documentation provided in the product datasheet and COA.