Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-oxo-1H-1,10-phenanthroline-3-carboxylic acid | 331830-20-7 | 98.4% | 1 ML
SDP

Catalog No. 5000231920
Encompass_Preferred

1,4-DPCA is a small-molecule prolyl-4-hydroxylase inhibitor supplied for research use. Available as solid material and as a ready-to-use 10 mM solution in DMSO (1 mL), it is used to inhibit collagen hydroxylation and to probe hypoxia-related pathways.

  • Ready-to-use 10 mM solution in DMSO (1 mL).
  • High purity suitable for research applications.
  • Molecular weight 240.21 g/mol; formula C13H8N2O3.
  • Soluble in DMSO; use freshly opened DMSO for best results.
  • Includes documented purity and solubility data for experimental planning.

Catalog No. 50-002-31920 Supplier Medchemexpress LLC Supplier No. HY12117210MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

1,4-DPCA is a small-molecule prolyl-4-hydroxylase inhibitor supplied for research use. Available as solid material and as a ready-to-use 10 mM solution in DMSO (1 mL), it is used to inhibit collagen hydroxylation and to probe hypoxia-related pathways.

  • Ready-to-use 10 mM solution in DMSO (1 mL).
  • High purity suitable for research applications.
  • Molecular weight 240.21 g/mol; formula C13H8N2O3.
  • Soluble in DMSO; use freshly opened DMSO for best results.
  • Includes documented purity and solubility data for experimental planning.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.