Learn More
Medchemexpress LLC 4-Oxo-2,2,6,6-tetramethylpiperidine-d16-1-15N-1-oxyl | 80404-14-4 | >98.0% | 187.32 g/mol | 15NC9D16O2 | 5 MG
SDP
Catalog No. 5000264897
Encompass_Preferred
Tempone-15N,d16 is an isotopically labeled nitroxide (15N and deuterium d16) derivative of Tempone provided for analytical and research use. It is used as a tracer and as an internal standard for quantitative analyses by NMR, GC-MS, and LC-MS. CAS 80404-14-4; molecular formula 15NC9D16O2; molecular weight 187.32 g/mol.
  • Isotopically labeled for unambiguous tracking in mass spectrometry.
  • Suitable as an internal standard for quantitative NMR, GC-MS, and LC-MS.
  • High isotopic enrichment and chemical purity (>98%).
  • Supplied in small, ready-to-use quantities for analytical workflows.
  • Stable nitroxide radical useful for spin-label and tracer applications.
Catalog No. 50-002-64897 Supplier Medchemexpress LLC Supplier No. HY129242S5MG
Tempone-15N,d16 is an isotopically labeled nitroxide (15N and deuterium d16) derivative of Tempone provided for analytical and research use. It is used as a tracer and as an internal standard for quantitative analyses by NMR, GC-MS, and LC-MS. CAS 80404-14-4; molecular formula 15NC9D16O2; molecular weight 187.32 g/mol.
  • Isotopically labeled for unambiguous tracking in mass spectrometry.
  • Suitable as an internal standard for quantitative NMR, GC-MS, and LC-MS.
  • High isotopic enrichment and chemical purity (>98%).
  • Supplied in small, ready-to-use quantities for analytical workflows.
  • Stable nitroxide radical useful for spin-label and tracer applications.