Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Oxo-3-quinazolinyl)acetic acid, 97%, Thermo Scientific™

Catalog No. AAH6672406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6672406 5 g
AAH6672403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6672406 Supplier Thermo Scientific Chemicals Supplier No. H6672406

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14663-53-7
Molecular Formula C10H8N2O3
Molecular Weight (g/mol) 204.185
MDL Number MFCD02332713
InChI Key BBBICQPTZBHLBM-UHFFFAOYSA-N
Synonym 4-oxo-4h-quinazolin-3-yl-acetic acid, 4-oxoquinazolin-3 4h-yl acetic acid, 2-4-oxoquinazolin-3 4h-yl acetic acid, 2-4-oxo-3,4-dihydroquinazolin-3-yl acetic acid, 4-oxoquinazolin-3-yl acetic acid, 3 4h-quinazolineacetic acid, 4-oxo, 4-oxo-3 4h-quinazolinyl acetic acid, 2-4-oxo-3-hydroquinazolin-3-yl acetic acid, 3 4h-quinazolineacetic acid,4-oxo, cambridge id 6340677
PubChem CID 729359
IUPAC Name 2-(4-oxoquinazolin-3-yl)acetic acid
SMILES C1=CC=C2C(=C1)C(=O)N(C=N2)CC(=O)O

Specifications

Quantity 5 g
Beilstein 184018
Formula Weight 204.18
Percent Purity 97%
Physical Form Solid
Chemical Name or Material (4-Oxo-3-quinazolinyl)acetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.