Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Oxo-Tempo

Catalog No. 1794855G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-948-55G 5 g
17-948-51G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 17-948-55G Supplier Sigma Aldrich Supplier No. 1794855G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Oxo-TEMPO is formed by the reaction of 2,2,6,6-tetramethyl-4-piperidone (4-oxo-TMP) and 1O2. It is a stable paramagnetic product formed during the irradiation of TiO2 in aqueous suspensions.Free-radical nitroxide spin probe typically used for:Redox sources for anodes in lithium secondary batteriesFree-radical biological studiesRadical spin-trappingElectron paramagnetic resonance studiesPolymer chemisty and synthesis applications

Specifications

CAS 2896-70-0
Molecular Formula C9H16NO2
Linear Formula C9H16NO2
MDL Number MFCD00006187
Quantity 5 g
Synonym 4-Oxo-2,2,6,6-tetramethyl-1-piperidinyloxy, free radical
Molecular Weight (g/mol) 170.23 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.