Learn More
Ace Glass, Inc. 4oz Stir-Lube stirrer lubrication
SDP
Catalog No. 509558286
Encompass_Preferred
4oz Stir-Lube stirrer lubricant is a superior, low melting, silicone based lubricant which liquefies at body temperature. Its composition allows for application of a very thin layer to a stirring shaft to increase bearing and shaft life at least three times over that of a bearing lubricated with glycerine.
Catalog No. 50-955-8286 Supplier Ace Glass, Inc. Supplier No. 811720
4oz Stir-Lube stirrer lubricant is a superior, low melting, silicone based lubricant which liquefies at body temperature. Its composition allows for application of a very thin layer to a stirring shaft to increase bearing and shaft life at least three times over that of a bearing lubricated with glycerine.