Learn More
Gilson Company Inc 4oz. Tinned-Metal Sample Container, 12/pk
Catalog No. NC0983301
Encompass_Preferred
4oz Tinned-Metal Sample Containers are 2.4x1.6in (61x41mm) Dia.xH and used for storing samples. Lids are included to protect samples from moisture changes. For use with HM-923 Vacuum Degassing Oven (VDO) and other testing processes. Sold in a package of 12.
Catalog No. NC0983301 Supplier Gilson Company Inc Supplier No. SC502
4oz Tinned-Metal Sample Containers are 2.4x1.6in (61x41mm) Dia.xH and used for storing samples. Lids are included to protect samples from moisture changes. For use with HM-923 Vacuum Degassing Oven (VDO) and other testing processes. Sold in a package of 12.