Learn More
O Berk Company 4OZ FLINT SAMPLE ROUND GLASS JAR - 48-400 144/Pk
Catalog No. NC9446061
Encompass_Preferred
GLASS JAR: 4OZ FLINT GLASS SAMPLE ROUND WITH 48-400NECK FINISH Size 4.00 oz; Neck Diameter48.00; Neck Finish 400; Material GLASS; Color FLINT; Shape Round 144/Pk
Catalog No. NC9446061 Supplier O Berk Company Supplier No. 28020236
GLASS JAR: 4OZ FLINT GLASS SAMPLE ROUND WITH 48-400NECK FINISH Size 4.00 oz; Neck Diameter48.00; Neck Finish 400; Material GLASS; Color FLINT; Shape Round 144/Pk