Learn More
Andwin Scientific 4 OZ SPECIMEN CONTAINER 100/CS
SDP
Catalog No. NC9547560
Encompass_Preferred
The 133 mL specimen container is made from sturdy polypropylene that can provide secure storage for most liquid and solid specimens. The product is translucent and is compatible to keep strong acids, bases, and most organic solvents. It also features graduations in both voirdupois and metric units. This product is sold in a case of 100 containers with polyethylene purple caps measuring 57.15 mm in diameter.
Catalog No. NC9547560 Supplier Andwin Scientific Supplier No. 25384146
Add to Cart
Add to Cart
The 133 mL specimen container is made from sturdy polypropylene that can provide secure storage for most liquid and solid specimens. The product is translucent and is compatible to keep strong acids, bases, and most organic solvents. It also features graduations in both voirdupois and metric units. This product is sold in a case of 100 containers with polyethylene purple caps measuring 57.15 mm in diameter.