Learn More
MSC Loctite 209681 4 oz Stick Two Part Epoxy 2.5 to 5 min Working Time, -30°C to 120°F, >500 psi Shear Strength
Catalog No. 501113848
Encompass
Loctite 209681 4 oz Stick Two Part Epoxy 2.5 to 5 min Working Time, -30°C to 120°F, >500 psi Shear Strength
Catalog No. 50-111-3848 Supplier MSC Supplier No. 05451877
Loctite 209681 4 oz Stick Two Part Epoxy 2.5 to 5 min Working Time, -30°C to 120°F, >500 psi Shear Strength