Learn More
Uline White Round Wide-Mouth Plastic Jars - 4 oz, White Cap, 36/cs
Catalog No. NC1626827
Encompass
Excellent for cosmetics, chemicals or nuts and bolts. Wide mouth, no inside lip for easy filling and pouring. Chemical-resistant polypropylene. FDA compliant. Caps included.
Catalog No. NC1626827 Supplier Uline Supplier No. S14506CS
Add to Cart
Add to Cart
Excellent for cosmetics, chemicals or nuts and bolts. Wide mouth, no inside lip for easy filling and pouring. Chemical-resistant polypropylene. FDA compliant. Caps included.