Learn More
TARGETMOL CHEMICALS INC 4-P-PDOT 10MG
SDP
Catalog No. 502880464
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 4-P-PDOT (4-phenyl-2- propionamidotetralin) is a potent, selective and affinity Melatonin receptor (MT2) antagonist. Purity 99.68%
Catalog No. 50-288-0464 Supplier TARGETMOL CHEMICALS INC Supplier No. T1404210MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 4-P-PDOT (4-phenyl-2- propionamidotetralin) is a potent, selective and affinity Melatonin receptor (MT2) antagonist. Purity 99.68%