Learn More
Medchemexpress LLC 4-p-pdot | 134865-74-0 | MFCD00901545 | >98.0% | 279.38 g/mol | C19H21NO | 25 MG
SDP
Catalog No. 5000213931
Encompass_Preferred
4-P-PDOT is a potent, selective melatonin MT2 receptor antagonist used in research to probe melatonin signaling and antioxidant pathways. It exhibits strong selectivity for the MT2 subtype and has been reported to counteract melatonin-mediated antioxidant responses such as changes in GSH/GSSG ratio and Nrf2 activity.
  • Potent, selective MT2 receptor antagonist.
  • Greater than 300-fold selectivity for MT2 versus MT1.
  • Useful for studies of melatonin-mediated antioxidant signaling.
  • Modulates GSH/GSSG ratio, phospho-ERK, and Nrf2 activity.
  • Available as an analytical standard for research use.
  • High purity (≥98%) suitable for biochemical assays.
Catalog No. 50-002-13931 Supplier Medchemexpress LLC Supplier No. HY10060925MG
4-P-PDOT is a potent, selective melatonin MT2 receptor antagonist used in research to probe melatonin signaling and antioxidant pathways. It exhibits strong selectivity for the MT2 subtype and has been reported to counteract melatonin-mediated antioxidant responses such as changes in GSH/GSSG ratio and Nrf2 activity.
  • Potent, selective MT2 receptor antagonist.
  • Greater than 300-fold selectivity for MT2 versus MT1.
  • Useful for studies of melatonin-mediated antioxidant signaling.
  • Modulates GSH/GSSG ratio, phospho-ERK, and Nrf2 activity.
  • Available as an analytical standard for research use.
  • High purity (≥98%) suitable for biochemical assays.