Learn More
Medchemexpress LLC 4-p-pdot | 134865-74-0 | MFCD00901545 | 99.5% | 279.4 g/mol | C19H21NO | 1 ML
SDP
Catalog No. 5000214713
Encompass_Preferred
4-P-PDOT is a selective melatonin MT2 receptor antagonist provided as a 10 mM solution in DMSO for research applications. It is used as an analytical reference compound in receptor pharmacology and neuroscience studies, and is accompanied by batch-specific documentation for safe handling and experimental reproducibility.
  • Selective MT2 receptor antagonist with high selectivity over MT1.
  • Supplied as a ready-to-use 10 mM solution in DMSO for convenience.
  • Suitable for receptor pharmacology and neurochemical research applications.
  • Accompanied by datasheet and safety data sheet for lot-specific information.
Catalog No. 50-002-14713 Supplier Medchemexpress LLC Supplier No. HY10060910MM 1ML
4-P-PDOT is a selective melatonin MT2 receptor antagonist provided as a 10 mM solution in DMSO for research applications. It is used as an analytical reference compound in receptor pharmacology and neuroscience studies, and is accompanied by batch-specific documentation for safe handling and experimental reproducibility.
  • Selective MT2 receptor antagonist with high selectivity over MT1.
  • Supplied as a ready-to-use 10 mM solution in DMSO for convenience.
  • Suitable for receptor pharmacology and neurochemical research applications.
  • Accompanied by datasheet and safety data sheet for lot-specific information.