Learn More
Medchemexpress LLC HY-100609 5mg Medchemexpress, 4-P-PDOT CAS:134865-74-0 Purity:>98%
SDP
Catalog No. 501967756
Encompass_Preferred
Medchemexpress, HY-100609 5mg 4-P-PDOT CAS:134865-74-0 4-P-PDOT is a potent, selective and affinity Melatonin receptor (MT2) antagonist. 4-P-PDOT is >300-fold more selective for MT2 than MT1. 4-P-PDOT significantly counteracts Melatonin-mediated antioxidant effects (GSH/GSSG ratio, phospho-ERK, Nrf2 nuclear translocation, Nrf2 DNA-binding activity)[3][4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7756 Supplier Medchemexpress LLC Supplier No. HY1006095MG
Medchemexpress, HY-100609 5mg 4-P-PDOT CAS:134865-74-0 4-P-PDOT is a potent, selective and affinity Melatonin receptor (MT2) antagonist. 4-P-PDOT is >300-fold more selective for MT2 than MT1. 4-P-PDOT significantly counteracts Melatonin-mediated antioxidant effects (GSH/GSSG ratio, phospho-ERK, Nrf2 nuclear translocation, Nrf2 DNA-binding activity)[3][4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.