Learn More
Medchemexpress LLC 4-(p-tolylthio)thieno[2,3-c]pyridine-2-carboxamide | 251992-66-2 | >98.0% | 300.39 g/mol | C15H12N2OS2 | 1 ML
SDP
Catalog No. 5000215800
Encompass_Preferred
A-205804 is a small-molecule research inhibitor of E-selectin and ICAM-1 expression with reported IC50 values of 20 nM and 25 nM, respectively. It has been described as an orally bioavailable lead compound and is supplied by research chemical vendors for in vitro and in vivo studies; verify formulation and handling information from the supplier certificate of analysis or SDS before use.
  • Potent inhibitor of E-selectin and ICAM-1 with low-nanomolar activity.
  • Reported orally bioavailable lead compound for in vivo evaluation.
  • Available as solid or DMSO solution for flexible assay preparation.
  • High reported purity suitable for research use (HPLC purity ≥98%).
  • Applicable for studies of inflammatory responses and cell adhesion regulation.
Catalog No. 50-002-15800 Supplier Medchemexpress LLC Supplier No. HY10022610MM 1ML
A-205804 is a small-molecule research inhibitor of E-selectin and ICAM-1 expression with reported IC50 values of 20 nM and 25 nM, respectively. It has been described as an orally bioavailable lead compound and is supplied by research chemical vendors for in vitro and in vivo studies; verify formulation and handling information from the supplier certificate of analysis or SDS before use.
  • Potent inhibitor of E-selectin and ICAM-1 with low-nanomolar activity.
  • Reported orally bioavailable lead compound for in vivo evaluation.
  • Available as solid or DMSO solution for flexible assay preparation.
  • High reported purity suitable for research use (HPLC purity ≥98%).
  • Applicable for studies of inflammatory responses and cell adhesion regulation.