Learn More
Cayman Chemical 4-palmItamIdo TEMPO 10mg
SDP
Catalog No. 502932814
Encompass_Preferred
A lipid-soluble spin label; incorporates into vesicles and cell membranes; has been used to study the entrapment of molecules within phosphatidylcholine vesicles
Catalog No. 50-293-2814 Supplier Cayman Chemical Supplier No. 2599510 MG
A lipid-soluble spin label; incorporates into vesicles and cell membranes; has been used to study the entrapment of molecules within phosphatidylcholine vesicles