Learn More
Electron Microscopy Sciences 4% Paraformaldehyde (formaldehyde) aqueous solution, 100 mL
SDP
Catalog No. 5025998
Encompass_Preferred
Paraformaldehyde - methanol free solution. A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives. 100 ml bottles.
Catalog No. 50-259-98 Supplier Electron Microscopy Sciences Supplier No. 1574100
Paraformaldehyde - methanol free solution. A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives. 100 ml bottles.