Learn More
Selleck Chemical LLC 4-PBA (Sodium Phenylbutyrate) S4125-10mM/1mL
SDP
Catalog No. 503109113
Encompass_Preferred
4-PBA (Sodium Phenylbutyrate) is a salt of 4-phenylbutyrate (4-PBA) or 4-phenylbutyric acid Sodium phenylbutyrate is a histone deacetylase inhibitor used to treat urea cycle disorders
Catalog No. 50-310-9113 Supplier Selleck Chemical LLC Supplier No. S412510MM/1ML
Add to Cart
Add to Cart
4-PBA (Sodium Phenylbutyrate) is a salt of 4-phenylbutyrate (4-PBA) or 4-phenylbutyric acid Sodium phenylbutyrate is a histone deacetylase inhibitor used to treat urea cycle disorders