Learn More
Medchemexpress LLC 4-Penten-1-ol | 821-09-0 | 99.99% | 86.13 | 100 G
SDP
Catalog No. 5000452342
Encompass_Preferred
4-Penten-1-ol is a biochemical reagent and flavor ingredient utilized across various products, including food, beverages, and perfumes. It functions as a versatile starting material for synthesizing organic compounds like pharmaceuticals and agrochemicals. Research has also explored its potential as a biobased solvent due to its low toxicity and biodegradability, and for its antimicrobial properties against certain bacteria and fungi, suggesting its utility in developing new antibacterial agents.
  • Used as a flavor ingredient in food, beverages, and perfumes.
  • Serves as a starting material for synthesizing organic compounds.
  • Investigated as a potential biobased solvent due to low toxicity and biodegradability.
  • Shows antimicrobial properties against certain bacteria and fungi.
Catalog No. 50-004-52342 Supplier Medchemexpress LLC Supplier No. HYW010639100G
4-Penten-1-ol is a biochemical reagent and flavor ingredient utilized across various products, including food, beverages, and perfumes. It functions as a versatile starting material for synthesizing organic compounds like pharmaceuticals and agrochemicals. Research has also explored its potential as a biobased solvent due to its low toxicity and biodegradability, and for its antimicrobial properties against certain bacteria and fungi, suggesting its utility in developing new antibacterial agents.
  • Used as a flavor ingredient in food, beverages, and perfumes.
  • Serves as a starting material for synthesizing organic compounds.
  • Investigated as a potential biobased solvent due to low toxicity and biodegradability.
  • Shows antimicrobial properties against certain bacteria and fungi.