Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Penten-1-yl Acetate 98%, Thermo Scientific™

Catalog No. AC149720500
Change view
Click to view available options
50g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC149720500 50g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC149720500 Supplier Thermo Scientific Chemicals Supplier No. 149720500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1576-85-8
CAS Min % 97.5
CAS Max % 100.0
Density 0.91g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H12O2
Refractive Index 1.4180 to 1.4200 (20°C, 589nm)
Linear Formula CH3CO2(CH2)3CH=CH2
Quantity 50g
MDL Number MFCD00008723
Beilstein 02, II, 151
Synonym 4-pentenyl acetate, 4-penten-1-yl acetate, 5-acetoxy-1-pentene, unii-v4sif0mabv, pent-4-en-1-yl acetate, pent-4-ene-1-yl acetate, ch3c o o ch2 3ch=ch2, v4sif0mabv, 4-penten-1-ol acetate, 4-pentenyl acetate #
InChI Key LVHDNIMNOMRZMF-UHFFFAOYSA-N
SMILES CC(=O)OCCCC=C
IUPAC Name pent-4-en-1-yl acetate
Molecular Weight (g/mol) 128.17
PubChem CID 74096
ChEBI CHEBI:87515
Formula Weight 128.17
Percent Purity ≥97.5%
Chemical Name or Material 4-Penten-1-yl acetate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-413-6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.