Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Penten-2-ol

Catalog No. P88075G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P88075G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P88075G Supplier Sigma Aldrich Supplier No. P88075G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Trans-2-hexen-1-ol undergoes allylic epoxidation to yield (2r,3r)-(+)-3-propyloxiranemethanol in high pressure carbon dioxide.trans-2-hexen-1-ol was used to evaluate the quality of protected designation of virgin olive oils by headspace solid-phase microextraction-gas chromatography using flame ionization detection and multivariate analysis
  • It was also used in encapsulation of vanadium catalysts in inorganic and hybrid matrices using sol-gel method

Specifications

CAS 625-31-0
Density 0.837 g/mL (at 25°C (literature))
Boiling Point 115°C to 116°C (lit.)
Molecular Formula C5H10O
Refractive Index n20/D 1.424 (literature)
Linear Formula H2C=CHCH2CH(OH)CH3
MDL Number MFCD00004556
Quantity 25 g
Synonym Allyl methyl carbinol
Molecular Weight (g/mol) 86.13
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.