Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Penten-2-ol, 96%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
625-31-0
C5H10O
86.13
MFCD00004556
ZHZCYWWNFQUZOR-UHFFFAOYSA-N
4-penten-2-ol,1-penten-4-ol,4-hydroxypent-1-ene,penten-4-ol,acmc-20apgd,acmc-1bael,4-emtem-2-ol,ch2=chch2ch oh ch3,zhzcywwnfquzor-uhfffaoysa
12247
pent-4-en-2-ol
CC(CC=C)O
625-31-0
C5H10O
86.13
MFCD00004556
ZHZCYWWNFQUZOR-UHFFFAOYSA-N
4-penten-2-ol,1-penten-4-ol,4-hydroxypent-1-ene,penten-4-ol,acmc-20apgd,acmc-1bael,4-emtem-2-ol,ch2=chch2ch oh ch3,zhzcywwnfquzor-uhfffaoysa
12247
pent-4-en-2-ol
CC(CC=C)O
Color Colorless
Density 0.8370g/mL
Quantity 5g
Refractive Index 1.4230 to 1.4250
Specific Gravity 0.837
Flash Point 25°C
Infrared Spectrum Authentic
Boiling Point 115.0°C to 116.0°C
Packaging Glass bottle
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Linear Formula H2C=CHCH2CH(OH)CH3
Solubility Information Solubility in water: fully miscible
Formula Weight 86.13
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 4-Penten-2-ol
Show More Show Less

GHS H Statement
Flammable liquid and vapor.

GHS P Statement
Store in a well-ventilated place.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Warning

EINECSNumber : 210-887-8