Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Penten-2-ol 99%, Thermo Scientific™

Catalog No. AC130020250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC130020250 25 g
AC130020050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC130020250 Supplier Thermo Scientific Chemicals Supplier No. 130020250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-31-0
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
InChI Key ZHZCYWWNFQUZOR-UHFFFAOYSA-N
Synonym 4-penten-2-ol, 1-penten-4-ol, 4-hydroxypent-1-ene, penten-4-ol, acmc-20apgd, acmc-1bael, 4-emtem-2-ol, ch2=chch2ch oh ch3, zhzcywwnfquzor-uhfffaoysa
PubChem CID 12247
IUPAC Name pent-4-en-2-ol
SMILES CC(CC=C)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.837
Infrared Spectrum Authentic
Refractive Index 1.4230 to 1.4250 (20°C, 589nm)
Linear Formula H2C=CHCH2CH(OH)CH3
Quantity 25 g
Beilstein 01, 443
Formula Weight 86.13
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 4-Penten-2-ol, 99%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement Flammable liquid and vapor.
Precautionary Statement GHS P Statement Store in a well-ventilated place. Keep container tightly closed. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
EINECSNumber 210-887-8
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.