Learn More
Medchemexpress LLC 4-Pentenoic acid, 2-amino-, (2S)- | 16338-48-0 | 98.0% | 115.13 | 10 G
SDP
Catalog No. 5000366991
Encompass_Preferred
L-Allylglycine is an amino acid derivative. It is an inhibitor for glutamate decarboxylase (GAD) that reduces GABA biosynthesis in the brain and exhibits convulsant activity. This product is for research use only.
  • Acts as an amino acid derivative
  • Inhibits glutamate decarboxylase (GAD)
  • Reduces GABA biosynthesis in the brain
  • Exhibits convulsant activity
Catalog No. 50-003-66991 Supplier Medchemexpress LLC Supplier No. HYW01290610G
L-Allylglycine is an amino acid derivative. It is an inhibitor for glutamate decarboxylase (GAD) that reduces GABA biosynthesis in the brain and exhibits convulsant activity. This product is for research use only.
  • Acts as an amino acid derivative
  • Inhibits glutamate decarboxylase (GAD)
  • Reduces GABA biosynthesis in the brain
  • Exhibits convulsant activity