Learn More
Medchemexpress LLC 4-pentenoic acid, 2-amino-, (2S)- | 16338-48-0 | ≥98.0% | 115.13 | 5 G
SDP
Catalog No. 5000454132
Encompass_Preferred
L-Allylglycine is an amino acid derivative that functions as an inhibitor for glutamate decarboxylase (GAD), thereby reducing GABA biosynthesis in the brain. It is known to exhibit convulsant activity.
  • Amino acid derivative
  • Inhibitor for glutamate decarboxylase (GAD)
  • Reduces GABA biosynthesis in the brain
  • Exhibits convulsant activity
  • Purity: ≥98.0%
  • For research use only
Catalog No. 50-004-54132 Supplier Medchemexpress LLC Supplier No. HYW0129065G
L-Allylglycine is an amino acid derivative that functions as an inhibitor for glutamate decarboxylase (GAD), thereby reducing GABA biosynthesis in the brain. It is known to exhibit convulsant activity.
  • Amino acid derivative
  • Inhibitor for glutamate decarboxylase (GAD)
  • Reduces GABA biosynthesis in the brain
  • Exhibits convulsant activity
  • Purity: ≥98.0%
  • For research use only