Learn More
Medchemexpress LLC 4-pentenoic acid | 591-80-0 | MFCD00004408 | 99.9% | 100.12 g/mol | C5H8O2 | 5g
SDP
Catalog No. 5000268863
Encompass_Preferred
4-Pentenoic acid is a medium-chain unsaturated fatty acid 4-Pentenoic acid has hypoglycemic and fatty acid oxidation inhibitory activities 4-Pentenoic acid can affect blood glucose metabolism and energy metabolism through mechanisms such as inhibiting long-chain fatty acid oxidation reducing gluconeogenesis and promoting glucose utilization[1][2]
Catalog No. 50-002-68863 Supplier Medchemexpress LLC Supplier No. HYY06245G
4-Pentenoic acid is a medium-chain unsaturated fatty acid 4-Pentenoic acid has hypoglycemic and fatty acid oxidation inhibitory activities 4-Pentenoic acid can affect blood glucose metabolism and energy metabolism through mechanisms such as inhibiting long-chain fatty acid oxidation reducing gluconeogenesis and promoting glucose utilization[1][2]