Learn More
Selleck Chemical LLC 4-Pentenoic acid
SDP
Catalog No. 5000003884
Encompass_Preferred
4-Pentenoic acid (Allylacetic acid 3-vinylpropionic acid 4-penten-1-oic acid) a flavouring ingredient is used to inhibit fatty acid oxidation in rat heart mitochondria
Catalog No. 50-000-03884 Supplier Selleck Chemical LLC Supplier No. S626825UL
4-Pentenoic acid (Allylacetic acid 3-vinylpropionic acid 4-penten-1-oic acid) a flavouring ingredient is used to inhibit fatty acid oxidation in rat heart mitochondria