Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Pentenoic acid, 98%

Catalog No. aaa1399109
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1399109 10 g
AAA1399114 25 g
AAA1399122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1399109 Supplier Thermo Scientific Chemicals Supplier No. A1399109
Only null left
Add to Cart

CAS: 591-80-0 | C5H8O2 | 100.12 g/mol

The sulfonation of 4-pentenoic acid (Allylacetic acid) yields the corresponding γ-lactone. It was used to inhibit fatty acid oxidation in rat heart mitochondria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The sulfonation of 4-pentenoic acid (Allylacetic acid) yields the corresponding γ-lactone. It was used to inhibit fatty acid oxidation in rat heart mitochondria.

Solubility
Soluble in alcohol, Slightly soluble in water

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-80-0
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00004408
InChI Key HVAMZGADVCBITI-UHFFFAOYSA-N
Synonym 4-pentenoic acid, allylacetic acid, allyl acetic acid, 4 pa, 3-vinylpropionic acid, delta 4-pentenoic acid, unii-d4s77y29fb, fema no. 2843, .delta.4-pentenoic acid, wln: qv3u1
PubChem CID 61138
ChEBI CHEBI:35936
IUPAC Name pent-4-enoic acid
SMILES OC(=O)CCC=C

Specifications

Melting Point -22°C
Density 0.891
Boiling Point 187°C to 189°C
Flash Point 89°C (192°F)
Odor Pungent
Refractive Index 1.4283
Quantity 10 g
UN Number UN3265
Beilstein 1633696
Solubility Information Soluble in alcohol, Slightly soluble in water
Formula Weight 100.12
Percent Purity 98%
Chemical Name or Material 4-Pentenoic acid
Show More Show Less
Hazard Category H227-H302-H314
Hazard Statement GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 209-732-7
RTECSNumber SB2800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.