Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-pentenyl isothiocyanate | 18060-79-2 | MFCD00191527 | 98.9% | 127.21 | C6H9NS | 25 MG
SDP

Catalog No. 5000281412
Encompass_Preferred

4-Pentenyl isothiocyanate is a naturally occurring isothiocyanate isolated from the aerial parts of Aurinia sinuata. Supplied for research use, the compound has molecular formula C6H9NS and molecular weight 127.21 g/mol. It is provided in small laboratory quantities with analytical documentation for verification.

  • Natural isothiocyanate from plant sources.
  • High purity: 98.88%.
  • Molecular formula C6H9NS; molecular weight 127.21 g/mol.
  • Appearance: colorless to light yellow liquid.
  • Storage recommendation: 4°C under nitrogen; in solvent -80°C (6 months), -20°C (1 month).
  • COA and SDS available for quality and safety verification.

Catalog No. 50-002-81412 Supplier Medchemexpress LLC Supplier No. HYW26979925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Pentenyl isothiocyanate is a naturally occurring isothiocyanate isolated from the aerial parts of Aurinia sinuata. Supplied for research use, the compound has molecular formula C6H9NS and molecular weight 127.21 g/mol. It is provided in small laboratory quantities with analytical documentation for verification.

  • Natural isothiocyanate from plant sources.
  • High purity: 98.88%.
  • Molecular formula C6H9NS; molecular weight 127.21 g/mol.
  • Appearance: colorless to light yellow liquid.
  • Storage recommendation: 4°C under nitrogen; in solvent -80°C (6 months), -20°C (1 month).
  • COA and SDS available for quality and safety verification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.