Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4'-Pentyl-4-biphenylcarbonitrile

Catalog No. 3285101G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-851-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-851-01G Supplier Sigma Aldrich Supplier No. 3285101G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Pentyl-4-biphenylcarbonitrile (5CB) is a nematic liquid crystal. 5CB undergoes phase transition from crystalline to nematic then to isotropic state at 18oC and 35oC respectively. It may be produced by modifying biphenyl.2Attaching 5CB to an aqueous-liquid crystal interface by redox active surfactant has been investigated.

Specifications

CAS 40817-08-1
Density 1.008 g/mL (at 25°C)
Boiling Point 140°C to 150°C (0.5 mmHg)
Molecular Formula C18H19N
Refractive Index n20/D 1.532 (literature)
Linear Formula CH3(CH2)4C6H4C6H4CN
MDL Number MFCD00036350
Quantity 1 g
Synonym 4-Cyano-4-pentylbiphenyl; 4-Pentyl-4-cyanobiphenyl; 5 CB
Molecular Weight (g/mol) 249.35
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.