Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Pentylbiphenyl 96.0+%, TCI America™
SDP

Catalog No. P11335ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11335ML 5 mL
P113325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11335ML Supplier TCI America Supplier No. P11335ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7116-96-3
Molecular Formula C17H20
Molecular Weight (g/mol) 224.35
MDL Number MFCD00009503
InChI Key IFUOTAQBVGAZPR-UHFFFAOYSA-N
Synonym 4-pentylbiphenyl, 4-n-pentylbiphenyl, 4-pentyl-1,1'-biphenyl, p-pentylbiphenyl, 1,1'-biphenyl, 4-pentyl, 4pentylbiphenyl, 1,1'-biphenyl, pentyl, 4-n-amyldiphenyl, 4-n-amylbiphenyl, pubchem12931
PubChem CID 81546
IUPAC Name 4-pentyl-1,1'-biphenyl
SMILES CCCCCC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 167°C
Quantity 5 mL
Formula Weight 224.35
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Pentylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.